Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%

$167.78 - $661.89

Chemical Identifiers

Chemical Name or Material (3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid
CAS 78266-06-5
Molecular Formula C15H19BrN2O5
Formula Weight 387.24
MDL Number MFCD00216974
Synonym Mebrofenin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1608603

 
alfa aesar™
L1608603
1g Each for $167.78

AAL1608606

 
alfa aesar™
L1608606
5g Each for $661.89
Description & Specifications

Specifications

Formula Weight 387.24
Assay ≥98%
CAS 78266-06-5
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid
Molecular Formula C15H19BrN2O5
MDL Number MFCD00216974
Melting Point 192° to 196°C
Synonym Mebrofenin
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number MB9121850

Applications
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.

Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.

Notes
Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.