missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1608603

 View more versions of this product

Catalog No. AAL1608603



Description

Description

(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.

Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid
192°C to 196°C
MFCD00216974
mebrofenin, choletec, mebrofenine inn-french, mebrofeninum inn-latin, mebrofenino inn-spanish, unii-7pv0b6ed98, 3-bromo-2,4,6-trimethylphenylcarbamoyl methyliminodiacetic acid, 3-bromomesityl carbamoyl methyl imino diacetic acid, n-2-3-bromo-2,4,6-trimethylphenyl amino-2-oxoethyl-n-carboxymethyl glycine, mebrofenine
MHPZZZZLAQGTHT-UHFFFAOYSA-N
2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid
54158
≥98%
78266-06-5
C15H19BrN2O5
1 g
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.
CC1=CC(=C(C(=C1NC(=O)CN(CC(=O)O)CC(=O)O)C)Br)C
387.23
387.24

Chemical Identifiers

78266-06-5
387.23
MHPZZZZLAQGTHT-UHFFFAOYSA-N
54158
CC1=CC(=C(C(=C1NC(=O)CN(CC(=O)O)CC(=O)O)C)Br)C
C15H19BrN2O5
MFCD00216974
mebrofenin, choletec, mebrofenine inn-french, mebrofeninum inn-latin, mebrofenino inn-spanish, unii-7pv0b6ed98, 3-bromo-2,4,6-trimethylphenylcarbamoyl methyliminodiacetic acid, 3-bromomesityl carbamoyl methyl imino diacetic acid, n-2-3-bromo-2,4,6-trimethylphenyl amino-2-oxoethyl-n-carboxymethyl glycine, mebrofenine
2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 278-877-6

RTECSNumber : MB9121850

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only