Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2,4,5-trifluorobenzoic acid, 98%

$70.19 - $914.34

Chemical Identifiers

Chemical Name or Material 3-Bromo-2,4,5-trifluorobenzoic acid
CAS 104222-42-6
Molecular Formula C7H2BrF3O2
Formula Weight 254.99
MDL Number MFCD05664065
Beilstein 7584381
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2015403

 
alfa aesar™
L2015403
1g Each for $70.19

AAL2015406

 
alfa aesar™
L2015406
5g Each for $235.51

AAL2015414

 
alfa aesar™
L2015414
25g Each for $914.34
Description & Specifications

Specifications

Formula Weight 254.99
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Bromo-2,4,5-trifluorobenzoic acid
Assay 98%
Molecular Formula C7H2BrF3O2
MDL Number MFCD05664065
CAS 104222-42-6
Health Hazard 1 Warning
Melting Point 122° to 123°C
Beilstein 7584381
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.