missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Bromo-2,2':5',2″-terthiophene 97.0+%, TCI America™
$807.14 - $3,526.79
Chemical Identifiers
| CAS | 105125-00-6 |
|---|---|
| Molecular Formula | C12H7BrS3 |
| Molecular Weight (g/mol) | 327.272 |
| InChI Key | FGBHDLKMGUOJBA-UHFFFAOYSA-N |
| Synonym | 3-Bromo-2,5-di(2-thienyl)thiophene |
| PubChem CID | 184442 |
| IUPAC Name | 3-bromo-2,5-dithiophen-2-ylthiophene |
| SMILES | C1=CSC(=C1)C2=CC(=C(S2)C3=CC=CS3)Br |
Chemical Identifiers
| 105125-00-6 | |
| 327.272 | |
| 3-Bromo-2,5-di(2-thienyl)thiophene | |
| 3-bromo-2,5-dithiophen-2-ylthiophene |
| C12H7BrS3 | |
| FGBHDLKMGUOJBA-UHFFFAOYSA-N | |
| 184442 | |
| C1=CSC(=C1)C2=CC(=C(S2)C3=CC=CS3)Br |
Specifications
| 105125-00-6 | |
| C12H7BrS3 | |
| 3-Bromo-2,5-di(2-thienyl)thiophene | |
| C1=CSC(=C1)C2=CC(=C(S2)C3=CC=CS3)Br | |
| 327.272 | |
| 327.27 | |
| 3′-Bromo-2,2′:5′,2′′-terthiophene |
| 40°C | |
| 1 g | |
| FGBHDLKMGUOJBA-UHFFFAOYSA-N | |
| 3-bromo-2,5-dithiophen-2-ylthiophene | |
| 184442 | |
| ≥97.0% (GC) |
Safety and Handling
TSCA : No