missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-1-trimethylsilyl-1-propyne, 98%

Manufacturer:  Alfa Aesar™ H5345106

 View more versions of this product

Catalog No. AAH5345106



Description

Description

Applications
3-Bromo-1-trimethylsilyl-1-propyne is used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It acts as a propargylating agent and used for the alkylation of dianion of beta-keto esters at gamma-carbon. It plays an important role in the synthesis of allenic alcohols and terminal conjugated enynes.

Solubility
Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

3-Bromo-1-trimethylsilyl-1-propyne
C6H11BrSi
MFCD00134460
3-bromo-1-propynyl trimethylsilane
GAPRPFRDVCCCHR-UHFFFAOYSA-N
3-bromoprop-1-ynyl(trimethyl)silane
642589
98%
1.483
5g
38002-45-8
BrCH2C≡CSi(CH3)3
2037650
Immiscible with water.
C[Si](C)(C)C#CCBr
191.143
191.14
1.17
44°C to 45°C (2mmHg)
63°C (145°F)

Chemical Identifiers

38002-45-8
191.143
GAPRPFRDVCCCHR-UHFFFAOYSA-N
642589
C[Si](C)(C)C#CCBr
C6H11BrSi
MFCD00134460
3-bromo-1-propynyl trimethylsilane
3-bromoprop-1-ynyl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents