missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-1-(phenylsulfonyl)indole, 97%

$37.78 - $148.81

Chemical Identifiers

CAS 99655-68-2
Molecular Formula C14H10BrNO2S
Molecular Weight (g/mol) 336.203
MDL Number MFCD02681986
InChI Key MXPFKHHDXIJNDX-UHFFFAOYSA-N
Synonym 3-bromo-1-phenylsulfonyl-1h-indole, 1-benzenesulfonyl-3-bromoindole, 3-bromo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-bromo-1h-indole, 1-phenylsulfonyl-3-bromoindole, 1-benzenesulphonyl-3-bromo-1h-indole, 3-bromo-1-phenylsulphonyl-1h-indole, 1-benzenesulfonyl-3-bromo-1h-indole, 3-bromo-1-benzenesulphonylindole, 1h-indole, 3-bromo-1-phenylsulfonyl
PubChem CID 2776215
IUPAC Name 1-(benzenesulfonyl)-3-bromoindole
SMILES C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6622603 View Documents Alfa Aesar™
H6622603
1g Each for $37.78
 
AAH6622606 View Documents Alfa Aesar™
H6622606
5g Each for $148.81
 
Specifications

Chemical Identifiers

99655-68-2
336.203
MXPFKHHDXIJNDX-UHFFFAOYSA-N
2776215
C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)Br
C14H10BrNO2S
MFCD02681986
3-bromo-1-phenylsulfonyl-1h-indole, 1-benzenesulfonyl-3-bromoindole, 3-bromo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-bromo-1h-indole, 1-phenylsulfonyl-3-bromoindole, 1-benzenesulphonyl-3-bromo-1h-indole, 3-bromo-1-phenylsulphonyl-1h-indole, 1-benzenesulfonyl-3-bromo-1h-indole, 3-bromo-1-benzenesulphonylindole, 1h-indole, 3-bromo-1-phenylsulfonyl
1-(benzenesulfonyl)-3-bromoindole

Specifications

122°C to 126°C
99655-68-2
MFCD02681986
3-bromo-1-phenylsulfonyl-1h-indole, 1-benzenesulfonyl-3-bromoindole, 3-bromo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-bromo-1h-indole, 1-phenylsulfonyl-3-bromoindole, 1-benzenesulphonyl-3-bromo-1h-indole, 3-bromo-1-phenylsulphonyl-1h-indole, 1-benzenesulfonyl-3-bromo-1h-indole, 3-bromo-1-benzenesulphonylindole, 1h-indole, 3-bromo-1-phenylsulfonyl
C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)Br
336.203
336.2
97%
1g
C14H10BrNO2S
3553533
MXPFKHHDXIJNDX-UHFFFAOYSA-N
1-(benzenesulfonyl)-3-bromoindole
2776215
Solid
3-Bromo-1-(phenylsulfonyl)indole
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning