Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-1-methylpyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromo-1-methylpyrrolidine
CAS 10603-45-9
Formula Weight 164.04
MDL Number MFCD12755877
UN Number UN2733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6625003

  SDS
alfa aesar™
H6625003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6625006

  SDS
alfa aesar™
H6625006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66250MD

  SDS
alfa aesar™
H66250MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.04
MDL Number MFCD12755877
Chemical Name or Material 3-Bromo-1-methylpyrrolidine
Health Hazard 1 Danger
CAS 10603-45-9
Physical Form Liquid
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Assay 97%
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Sensitivity Moisture Sensitive
Density 1.361g/mL
Refractive Index 1.4910
Storage Note 1 2° to 8°C
Color Yellow to brown
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2733
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

2° to 8°C