Please login to your online account to display your discounted pricing

3-Bromo-1-benzofuran, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC27510CB

 View more versions of this product

Catalog No. CC27510CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.03
Quantity 250mg
Assay 97%
CAS 59214-70-9
Assay Percent Range 97%
Molecular Formula C8H5BrO
Packaging Amber Glass Bottle
Chemical Name or Material 3-Bromo-1-benzofuran

Specifications

Chemical Name or Material 3-Bromo-1-benzofuran
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H5BrO
Formula Weight 197.03