Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-1,1,1-trifluoro-2-propanol, 96%

Chemical Identifiers

Chemical Name or Material 3-Bromo-1,1,1-trifluoro-2-propanol
CAS 431-34-5
Molecular Formula C3H4BrF3O
Formula Weight 192.97
MDL Number MFCD00040945
UN Number UN1987
Beilstein 1737803
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1056603

  SDS
alfa aesar™
L1056603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1056606

  SDS
alfa aesar™
L1056606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.97
CAS 431-34-5
Chemical Name or Material 3-Bromo-1,1,1-trifluoro-2-propanol
Assay 96%
Molecular Formula C3H4BrF3O
MDL Number MFCD00040945
Quantity 1g
Health Hazard 2 GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
UN Number UN1987
Beilstein 1737803
Boiling Point 124° to 126°C
Flash Point 43°C (109°F)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280h-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P261-P280h-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.