missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-[Boc(methyl)aminomethyl]azetidine, 95%, Thermo Scientific Chemicals
$380.04 - $1085.77
Chemical Identifiers
CAS | 1053655-53-0 |
---|---|
Molecular Formula | C10H20N2O2 |
Molecular Weight (g/mol) | 200.282 |
MDL Number | MFCD08061967 |
InChI Key | IIRJNOLNCXOXOR-UHFFFAOYSA-N |
Synonym | tert-butyl azetidin-3-ylmethyl methyl carbamate, 3-n-boc-methylaminomethyl azetidine, tert-butyl n-azetidin-3-ylmethyl-n-methylcarbamate, 3-boc methyl aminomethyl azetidine, tert-butyl azetidin-3-ylmethyl, azetidin-3-ylmethyl neopentyl carbamate, tert-butylazetidin-3-ylmethyl methyl carbamate, azetidin-3-ylmethyl-methyl-carbamic acid tert-buty |
PubChem CID | 37818419 |
IUPAC Name | tert-butyl N-(azetidin-3-ylmethyl)-N-methylcarbamate |
SMILES | CC(C)(C)OC(=O)N(C)CC1CNC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH60310MD
|
Thermo Scientific Chemicals
H60310MD |
250 mg |
Each for $380.04
|
|
|||||
AAH6031003
|
Thermo Scientific Chemicals
H6031003 |
1 g |
Each for $1,085.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1053655-53-0 | |
200.282 | |
IIRJNOLNCXOXOR-UHFFFAOYSA-N | |
37818419 | |
CC(C)(C)OC(=O)N(C)CC1CNC1 |
C10H20N2O2 | |
MFCD08061967 | |
tert-butyl azetidin-3-ylmethyl methyl carbamate, 3-n-boc-methylaminomethyl azetidine, tert-butyl n-azetidin-3-ylmethyl-n-methylcarbamate, 3-boc methyl aminomethyl azetidine, tert-butyl azetidin-3-ylmethyl, azetidin-3-ylmethyl neopentyl carbamate, tert-butylazetidin-3-ylmethyl methyl carbamate, azetidin-3-ylmethyl-methyl-carbamic acid tert-buty | |
tert-butyl N-(azetidin-3-ylmethyl)-N-methylcarbamate |
Specifications
1053655-53-0 | |
MFCD08061967 | |
IIRJNOLNCXOXOR-UHFFFAOYSA-N | |
tert-butyl N-(azetidin-3-ylmethyl)-N-methylcarbamate | |
37818419 | |
95% | |
3-[Boc(methyl)aminomethyl]azetidine |
C10H20N2O2 | |
tert-butyl azetidin-3-ylmethyl methyl carbamate, 3-n-boc-methylaminomethyl azetidine, tert-butyl n-azetidin-3-ylmethyl-n-methylcarbamate, 3-boc methyl aminomethyl azetidine, tert-butyl azetidin-3-ylmethyl, azetidin-3-ylmethyl neopentyl carbamate, tert-butylazetidin-3-ylmethyl methyl carbamate, azetidin-3-ylmethyl-methyl-carbamic acid tert-buty | |
CC(C)(C)OC(=O)N(C)CC1CNC1 | |
200.282 | |
200.28 | |
250 mg |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only