missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-3-(Boc-aminomethyl)piperidine, 98%

$70.47 - $220.53

Chemical Identifiers

CAS 142643-29-6
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01632454
InChI Key KHPQHXGYYXYTDN-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethylpiperidine, tert-butyl piperidin-3-ylmethyl carbamate, 3-boc-aminomethyl-piperidine, 3-n-boc-aminomethyl piperidine, 3-boc-aminomethylpiperidine, tert-butyl n-piperidin-3-ylmethyl carbamate, piperidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidylmethyl carbamate, tert-butyl piperidin-3-ylmethylcarbamate, tert-butyl piperidin-3-yl methylcarbamate
PubChem CID 2756030
IUPAC Name tert-butyl N-(piperidin-3-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1CCCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60257MD View Documents Alfa Aesar™
H60257MD
250mg Each for $70.47
 
AAH6025703 View Documents Alfa Aesar™
H6025703
1g Each for $220.53
 
Specifications

Chemical Identifiers

142643-29-6
214.309
KHPQHXGYYXYTDN-UHFFFAOYSA-N
2756030
CC(C)(C)OC(=O)NCC1CCCNC1
C11H22N2O2
MFCD01632454
3-n-boc-aminomethylpiperidine, tert-butyl piperidin-3-ylmethyl carbamate, 3-boc-aminomethyl-piperidine, 3-n-boc-aminomethyl piperidine, 3-boc-aminomethylpiperidine, tert-butyl n-piperidin-3-ylmethyl carbamate, piperidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidylmethyl carbamate, tert-butyl piperidin-3-ylmethylcarbamate, tert-butyl piperidin-3-yl methylcarbamate
tert-butyl N-(piperidin-3-ylmethyl)carbamate

Specifications

60°C to 70°C
142643-29-6
MFCD01632454
7023643
KHPQHXGYYXYTDN-UHFFFAOYSA-N
tert-butyl N-(piperidin-3-ylmethyl)carbamate
2756030
98%
250mg
C11H22N2O2
UN3259
3-n-boc-aminomethylpiperidine, tert-butyl piperidin-3-ylmethyl carbamate, 3-boc-aminomethyl-piperidine, 3-n-boc-aminomethyl piperidine, 3-boc-aminomethylpiperidine, tert-butyl n-piperidin-3-ylmethyl carbamate, piperidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidylmethyl carbamate, tert-butyl piperidin-3-ylmethylcarbamate, tert-butyl piperidin-3-yl methylcarbamate
CC(C)(C)OC(=O)NCC1CCCNC1
214.309
214.3
(+/-)-3-(Boc-aminomethyl)piperidine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

Recommended Storage : 2° to 8°C