Please login to your online account to display your discounted pricing

3-(BOC-Aminomethyl)benzoic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(BOC-Aminomethyl)benzoic acid
CAS 117445-22-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H17NO4
Formula Weight 251.28
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC437870050

 
thermo fisher scientific
AC437870050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC437870010

 
thermo fisher scientific
AC437870010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Flakes
Assay ≥96.0%
Formula Weight 251.28
Quantity 5g
Color White to off-white
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-(BOC-Aminomethyl)benzoic acid
Assay Percent Range 97%
Molecular Formula C13H17NO4
CAS 117445-22-4
Infrared Spectrum Authentic