missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(BOC-Aminomethyl)benzoic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437870010
Specifications
3-(BOC-Aminomethyl)benzoic acid | |
Glass bottle | |
129.0°C to 134.8°C | |
117445-22-4 | |
100.0 | |
C13H17NO4 | |
MQNHKLMHRZYTBZ-UHFFFAOYSA-N | |
3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid | |
2755929 | |
Crystalline Powder or Flakes |
Authentic | |
White | |
1g | |
96.0 | |
96% min. (HPLC) | |
3-boc-aminomethyl benzoic acid, 3-boc-aminomethyl-benzoic acid, 3-n-boc-aminomethyl benzoic acid, 3-tert-butoxycarbonyl amino methyl benzoic acid, boc-3-amb-oh, boc-3-aminomethylbenzoic acid, boc-mamb, n-boc-3-aminomethyl benzoic acid, n-boc-3-aminomethyl-benzoic acid, n-boc-3-aminomethylbenzoic acid | |
CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C(=O)O | |
251.28 | |
251.28 | |
97% |
Chemical Identifiers
117445-22-4 | |
251.28 | |
3-boc-aminomethyl benzoic acid, 3-boc-aminomethyl-benzoic acid, 3-n-boc-aminomethyl benzoic acid, 3-tert-butoxycarbonyl amino methyl benzoic acid, boc-3-amb-oh, boc-3-aminomethylbenzoic acid, boc-mamb, n-boc-3-aminomethyl benzoic acid, n-boc-3-aminomethyl-benzoic acid, n-boc-3-aminomethylbenzoic acid | |
3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid |
C13H17NO4 | |
MQNHKLMHRZYTBZ-UHFFFAOYSA-N | |
2755929 | |
CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C(=O)O |