missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Boc-aminomethyl)azetidine hydrochloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H59816ME

 View more versions of this product

Catalog No. AAH59816ME



Description

Description

It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

3-(Boc-aminomethyl)azetidine hydrochloride
C9H18N2O2
3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine
MOLUHRBHXXGWDP-UHFFFAOYSA-N
tert-butyl N-[(azetidin-3-yl)methyl]carbamate
15346872
95%
91188-15-7
MFCD01861760
Slightly soluble in water.
CC(C)(C)OC(=O)NCC1CNC1
186.26
222.71
500 mg

Chemical Identifiers

91188-15-7
186.26
MOLUHRBHXXGWDP-UHFFFAOYSA-N
15346872
C9H18N2O2
MFCD01861760
3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine
CC(C)(C)OC(=O)NCC1CNC1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only