missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester, 96%

$242.69 - $728.07

Chemical Identifiers

CAS 1035391-48-0
Molecular Formula C18H27BFNO4
Molecular Weight (g/mol) 351.225
InChI Key VAYYEUXFQWTMJJ-UHFFFAOYSA-N
Synonym tert-butyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate
PubChem CID 59279260
IUPAC Name tert-butyl N-[[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)CNC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62453MD View Documents Alfa Aesar™
H62453MD
250mg Each for $242.69
 
AAH6245303 View Documents Alfa Aesar™
H6245303
1g Each for $728.07
 
Specifications

Chemical Identifiers

1035391-48-0
351.225
tert-butyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate
tert-butyl N-[[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
C18H27BFNO4
VAYYEUXFQWTMJJ-UHFFFAOYSA-N
59279260
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)CNC(=O)OC(C)(C)C

Specifications

250mg
C18H27BFNO4
VAYYEUXFQWTMJJ-UHFFFAOYSA-N
tert-butyl N-[[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
59279260
Solid
3-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester
1035391-48-0
tert-butyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)CNC(=O)OC(C)(C)C
351.225
351.22
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures