missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62453MD

 View more versions of this product

Catalog No. AAH62453MD



Specifications

Specifications

3-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester
1035391-48-0
tert-butyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)CNC(=O)OC(C)(C)C
351.225
351.22
96%
250mg
C18H27BFNO4
VAYYEUXFQWTMJJ-UHFFFAOYSA-N
tert-butyl N-[[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
59279260
Solid

Chemical Identifiers

1035391-48-0
351.225
tert-butyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate
tert-butyl N-[[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
C18H27BFNO4
VAYYEUXFQWTMJJ-UHFFFAOYSA-N
59279260
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)CNC(=O)OC(C)(C)C
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents