missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(BOC-amino)pyrrolidine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 452320010
Specifications
3-(BOC-amino)pyrrolidine | |
96% min. (GC) | |
C9H18N2O2 | |
3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine | |
CC(C)(C)OC(=O)NC1CCNC1 | |
186.26 | |
186.26 |
99724-19-3 | |
Glass Bottle | |
1g | |
DQQJBEAXSOOCPG-UHFFFAOYSA-N | |
tert-butyl N-pyrrolidin-3-ylcarbamate | |
2757234 | |
97% |
Chemical Identifiers
99724-19-3 | |
186.26 | |
3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine | |
tert-butyl N-pyrrolidin-3-ylcarbamate |
C9H18N2O2 | |
DQQJBEAXSOOCPG-UHFFFAOYSA-N | |
2757234 | |
CC(C)(C)OC(=O)NC1CCNC1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO â Research Use Only