Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-amino)cyclohexanone, 95%

Chemical Identifiers

Chemical Name or Material 3-(Boc-amino)cyclohexanone
CAS 885280-38-6
Formula Weight 213.28
MDL Number MFCD03790905
Beilstein 11040732
Synonym tert-Butyl 3-oxocyclohexylcarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6659403

 
alfa aesar™
H6659403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6659406

 
alfa aesar™
H6659406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66594MD

 
alfa aesar™
H66594MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.28
MDL Number MFCD03790905
Chemical Name or Material 3-(Boc-amino)cyclohexanone
CAS 885280-38-6
Beilstein 11040732
Physical Form Solid
Quantity 1g
Storage Note 1 -30° to -10°C
Synonym tert-Butyl 3-oxocyclohexylcarbamate
Assay 95%
Safety and Handling

-30° to -10°C