missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Boc-amino)cyclohexanone, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6659403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(Boc-amino)cyclohexanone | |
C11H19NO3 | |
11040732 | |
VGDCXKATFLOEHF-UHFFFAOYNA-N | |
tert-butyl N-(3-oxocyclohexyl)carbamate | |
44228975 | |
95% | |
Solid |
885280-38-6 | |
MFCD03790905 | |
3-n-boc-aminocyclohexanone, tert-butyl 3-oxocyclohexylcarbamate, 3-oxo-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl 3-oxocyclohexyl carbamate, 3-boc-amino cyclohexanone, tert-butyl n-3-oxocyclohexyl carbamate, tert-butyl3-oxocyclohexylcarbamate, pubchem20066, 3-n-boc-amino cyclohexanone, 3-boc-amino cyclohexan-1-one | |
CC(C)(C)OC(=O)NC1CCCC(=O)C1 | |
213.28 | |
213.28 | |
1 g |
Chemical Identifiers
885280-38-6 | |
213.28 | |
VGDCXKATFLOEHF-UHFFFAOYNA-N | |
44228975 | |
CC(C)(C)OC(=O)NC1CCCC(=O)C1 |
C11H19NO3 | |
MFCD03790905 | |
3-n-boc-aminocyclohexanone, tert-butyl 3-oxocyclohexylcarbamate, 3-oxo-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl 3-oxocyclohexyl carbamate, 3-boc-amino cyclohexanone, tert-butyl n-3-oxocyclohexyl carbamate, tert-butyl3-oxocyclohexylcarbamate, pubchem20066, 3-n-boc-amino cyclohexanone, 3-boc-amino cyclohexan-1-one | |
tert-butyl N-(3-oxocyclohexyl)carbamate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only