missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-amino)-3-(4-fluorophenyl)propionic acid, 98+%

$65.57 - $200.11

Chemical Identifiers

CAS 284493-72-7
Molecular Formula C14H18FNO4
Formula Weight 283.3
MDL Number MFCD02090715
Synonym 3-(tert-Butoxycarbonylamino)-3-(4-fluorophenyl)propionic acid
Chemical Name or Material 3-(Boc-amino)-3-(4-fluorophenyl)propionic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1980603 SDS Alfa Aesar™
L1980603
1g Each for $200.11
AAL19806MD SDS Alfa Aesar™
L19806MD
250mg Each for $65.57
Specifications Description & Specifications

Chemical Identifiers

284493-72-7
283.3
3-(tert-Butoxycarbonylamino)-3-(4-fluorophenyl)propionic acid
C14H18FNO4
MFCD02090715
3-(Boc-amino)-3-(4-fluorophenyl)propionic acid

Specifications

284493-72-7
152° to 154°C
MFCD02090715
3-(tert-Butoxycarbonylamino)-3-(4-fluorophenyl)propionic acid
≥98%
C14H18FNO4
283.3
1g
3-(Boc-amino)-3-(4-fluorophenyl)propionic acid

Safety and Handling

Recommended Storage : 2° to 8°C