Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-amino)-3-(4-chlorophenyl)propionic acid, 97%

$79.27 - $240.12

Chemical Identifiers

Chemical Name or Material 3-(Boc-amino)-3-(4-chlorophenyl)propionic acid
CAS 284493-65-8
Molecular Formula C14H18ClNO4
Formula Weight 299.75
MDL Number MFCD02090708
Synonym 3-(tert-Butoxycarbonylamino)-3-(4-chlorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1980303

 
alfa aesar™
L1980303
1g Each for $240.12

AAL19803MD

 
alfa aesar™
L19803MD
250mg Each for $79.27
Description & Specifications

Specifications

Formula Weight 299.75
Chemical Name or Material 3-(Boc-amino)-3-(4-chlorophenyl)propionic acid
Assay 97%
Molecular Formula C14H18ClNO4
MDL Number MFCD02090708
CAS 284493-65-8
Synonym 3-(tert-Butoxycarbonylamino)-3-(4-chlorophenyl)propionic acid
Recommended Storage 2° to 8°C
Melting Point 179° to 180°C
Quantity 1g
Safety and Handling

2° to 8°C