Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, 99%

$77.59 - $235.05

Chemical Identifiers

Chemical Name or Material 3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
CAS 284493-53-4
Molecular Formula C15H21NO5
Formula Weight 295.34
MDL Number MFCD02090693
Synonym 3-(tert-Butoxycarbonylamino)-3-(3-methoxyphenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1980703

  SDS
alfa aesar™
L1980703
1g Each for $235.05

AAL19807MD

  SDS
alfa aesar™
L19807MD
250mg Each for $77.59
Description & Specifications

Specifications

Formula Weight 295.34
CAS 284493-53-4
Chemical Name or Material 3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Molecular Formula C15H21NO5
Assay 99%
MDL Number MFCD02090693
Recommended Storage 2° to 8°C
Quantity 1g
Synonym 3-(tert-Butoxycarbonylamino)-3-(3-methoxyphenyl)propionic acid
Melting Point 81° to 83°C
Safety and Handling

2° to 8°C