Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-amino)-3-(3-fluorophenyl)propionic acid, 99%

$218.49 - $218.49

Chemical Identifiers

Chemical Name or Material 3-(Boc-amino)-3-(3-fluorophenyl)propionic acid
CAS 284493-59-0
Molecular Formula C14H18FNO4
Formula Weight 283.3
MDL Number MFCD02090701
Synonym 3-(tert-Butoxycarbonylamino)-3-(3-fluorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1980503

  SDS
alfa aesar™
L1980503
1g Each for $218.49
Description & Specifications

Specifications

Formula Weight 283.3
Chemical Name or Material 3-(Boc-amino)-3-(3-fluorophenyl)propionic acid
Assay 99%
CAS 284493-59-0
Molecular Formula C14H18FNO4
Synonym 3-(tert-Butoxycarbonylamino)-3-(3-fluorophenyl)propionic acid
Quantity 1g
Melting Point 124° to 126°C
MDL Number MFCD02090701