Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Boc-amino)-3-(2-fluorophenyl)propionic acid, 98+%

$200.11 - $200.11

Chemical Identifiers

Chemical Name or Material 3-(Boc-amino)-3-(2-fluorophenyl)propionic acid
CAS 284493-56-7
Molecular Formula C14H18FNO4
Formula Weight 283.3
MDL Number MFCD02090698
Synonym 3-(tert-Butoxycarbonylamino)-3-(2-fluorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1980403

  SDS
alfa aesar™
L1980403
1g Each for $200.11
Description & Specifications

Specifications

Formula Weight 283.3
Synonym 3-(tert-Butoxycarbonylamino)-3-(2-fluorophenyl)propionic acid
Chemical Name or Material 3-(Boc-amino)-3-(2-fluorophenyl)propionic acid
Assay ≥98%
Molecular Formula C14H18FNO4
MDL Number MFCD02090698
CAS 284493-56-7
Melting Point 142° to 144°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C