missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(BOC-amino)-1-propanol, 97%, ACROS Organics™

Chemical Identifiers

CAS 58885-58-8
Assay Percent Range 97%
Molecular Formula C8H17NO3
Linear Formula HO(CH2)3NHCO2C(CH3)3
MDL Number MFCD00191883
Synonym N-(3-Hydroxypropyl)carbamic acid tert-butyl ester
Chemical Name or Material 3-(BOC-amino)-1-propanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC399800100 SDS Thermo Fisher Scientific
AC399800100
10mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399800500 SDS Thermo Fisher Scientific
AC399800500
50mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

58885-58-8
C8H17NO3
MFCD00191883
3-(BOC-amino)-1-propanol
97%
HO(CH2)3NHCO2C(CH3)3
N-(3-Hydroxypropyl)carbamic acid tert-butyl ester

Specifications

58885-58-8
C8H17NO3
MFCD00191883
N-(3-Hydroxypropyl)carbamic acid tert-butyl ester
97%
HO(CH2)3NHCO2C(CH3)3
10mL
3-(BOC-amino)-1-propanol