Please login to your online account to display your discounted pricing

3α-Bis-(4-fluorophenyl) methoxytropane hydrochloride, Tocris


Potent dopamine uptake inhibitor

Quantity 50mg

Manufacturer: r & d systems  091850

 View more versions of this product

Catalog No. 091850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 379.88
CAS 202646-03-5
Molecular Formula C21H23F2NO·HCl
Recommended Storage Store at Room Temperature
Chemical Name or Material 3α-Bis-(4-fluorophenyl) methoxytropane hydrochloride
Assay Percent Range >98%

Specifications

Chemical Name or Material 3α-Bis-(4-fluorophenyl) methoxytropane hydrochloride
Assay Percent Range >98%
Molecular Formula C21H23F2NO·HCl
Formula Weight 379.88
Accessories

Store at Room Temperature