Please login to your online account to display your discounted pricing

3α-Bis-(4-fluorophenyl) methoxytropane hydrochloride, Tocris

Potent dopamine uptake inhibitor

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material 3α-Bis-(4-fluorophenyl) methoxytropane hydrochloride
Assay Percent Range >98%
Molecular Formula C21H23F2NO·HCl
Formula Weight 379.88
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

091810

 
r & d systems
091810
10mg Each for $133.98

091850

 
r & d systems
091850
50mg Each for $567.83
Description & Specifications

Specifications

Chemical Name or Material 3α-Bis-(4-fluorophenyl) methoxytropane hydrochloride
CAS 202646-03-5
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C21H23F2NO·HCl
Formula Weight 379.88
Quantity 10mg
Safety and Handling

Store at Room Temperature