missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid, 99%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5618730

 View more versions of this product

Catalog No. AAH5618730



Specifications

Specifications

3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid
68399-80-4
MFCD00038353
1-propanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy
C(CO)N(CCO)CC(CS(=O)(=O)O)O
243.274
261.29
250g
C7H17NO6S
2105454
XCBLFURAFHFFJF-UHFFFAOYSA-N
3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
100210
99%

Chemical Identifiers

68399-80-4
243.274
XCBLFURAFHFFJF-UHFFFAOYSA-N
100210
C(CO)N(CCO)CC(CS(=O)(=O)O)O
C7H17NO6S
MFCD00038353
1-propanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy
3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
Safety and Handling

Safety and Handling

EINECSNumber : 269-992-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents