missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%

Manufacturer:  Alfa Aesar™ H5575403

 View more versions of this product

Catalog No. AAH5575403



Specifications

Specifications

(3-Benzyloxypropyl)triphenylphosphonium bromide
153°C to 154°C
54314-85-1
MFCD00191781
DWYCJWXMZGVGJV-UHFFFAOYSA-M
triphenyl(3-phenylmethoxypropyl)phosphanium;bromide
10929077
Powder
White
1g
C28H28BrOP
3-benzyloxy propyl triphenylphosphonium bromide
C1=CC=C(C=C1)COCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
491.409
491.41
98%

Chemical Identifiers

54314-85-1
491.409
DWYCJWXMZGVGJV-UHFFFAOYSA-M
10929077
C1=CC=C(C=C1)COCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C28H28BrOP
MFCD00191781
3-benzyloxy propyl triphenylphosphonium bromide
triphenyl(3-phenylmethoxypropyl)phosphanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents