missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%

Chemical Identifiers

CAS 54314-85-1
Formula Weight 491.41
MDL Number MFCD00191781
Chemical Name or Material (3-Benzyloxypropyl)triphenylphosphonium bromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5575403 SDS Alfa Aesar™
H5575403
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5575406 SDS Alfa Aesar™
H5575406
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5575414 SDS Alfa Aesar™
H5575414
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

54314-85-1
491.41
MFCD00191781
(3-Benzyloxypropyl)triphenylphosphonium bromide

Specifications

54314-85-1
491.41
MFCD00191781
1g
98%
Powder
153° to 154°C
White to off-white
(3-Benzyloxypropyl)triphenylphosphonium bromide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning