missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Benzyloxypropyl)triphenylphosphonium bromide, ≥97%

Manufacturer:  Alfa Aesar™ H5575406

 View more versions of this product

Catalog No. AAH5575406



Specifications

Specifications

(3-Benzyloxypropyl)triphenylphosphonium bromide
153°C to 154°C
54314-85-1
MFCD00191781
DWYCJWXMZGVGJV-UHFFFAOYSA-M
triphenyl(3-phenylmethoxypropyl)phosphanium;bromide
10929077
Powder
White
5g
C28H28BrOP
3-benzyloxy propyl triphenylphosphonium bromide
C1=CC=C(C=C1)COCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
491.409
491.41
98%

Chemical Identifiers

54314-85-1
491.409
DWYCJWXMZGVGJV-UHFFFAOYSA-M
10929077
C1=CC=C(C=C1)COCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C28H28BrOP
MFCD00191781
3-benzyloxy propyl triphenylphosphonium bromide
triphenyl(3-phenylmethoxypropyl)phosphanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents