Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxyphenylacetonitrile, 97%

$207.93 - $207.93

Chemical Identifiers

Chemical Name or Material 3-Benzyloxyphenylacetonitrile
CAS 20967-96-8
Molecular Formula C15H13NO
Formula Weight 223.28
MDL Number MFCD00016396
Beilstein 2585802
Synonym 3-Benzyloxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1924104

  SDS
alfa aesar™
L1924104
2g Each for $207.93
Description & Specifications

Specifications

Formula Weight 223.28
CAS 20967-96-8
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
Molecular Formula C15H13NO
Melting Point 47° to 50°C
Chemical Name or Material 3-Benzyloxyphenylacetonitrile
Assay 97%
MDL Number MFCD00016396
Synonym 3-Benzyloxybenzyl cyanide
Quantity 2g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Boiling Point 188° to 190°C (3mmHg)
Beilstein 2585802
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.