missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzyloxybenzyl alcohol, 97%, ACROS Organics™

$146.72 - $146.72

Chemical Identifiers

CAS 1700-30-7
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00004642
InChI Key AFKLSWIRJUJWKY-UHFFFAOYSA-N
Synonym 3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
PubChem CID 74341
IUPAC Name (3-phenylmethoxyphenyl)methanol
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC181940010 SDS View Documents Acros Organics
AC181940010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC181940100 SDS View Documents Acros Organics
181940100
10g Glass bottle Each for $146.72
 
Specifications

Chemical Identifiers

1700-30-7
214.264
AFKLSWIRJUJWKY-UHFFFAOYSA-N
74341
C1=CC=C(C=C1)COC2=CC=CC(=C2)CO
C14H14O2
MFCD00004642
3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
(3-phenylmethoxyphenyl)methanol

Specifications

>110°C
Glass bottle
47.0°C to 52.0°C
1700-30-7
100.0
C14H14O2
MFCD00004642
AFKLSWIRJUJWKY-UHFFFAOYSA-N
(3-phenylmethoxyphenyl)methanol
74341
Powder
3-Benzyloxybenzyl alcohol
Authentic
Beige to Yellow
1g
96.0
97%
C6H5CH2OC6H4CH2OH
3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
C1=CC=C(C=C1)COC2=CC=CC(=C2)CO
214.264
214.26
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.