Please login to your online account to display your discounted pricing

3-Benzyloxybenzyl alcohol, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  181940100

 View more versions of this product

Catalog No. AC181940100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 214.26
Quantity 10g
Color Light yellow to light beige
Melting Point 47° to 52°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C14H14O2
Chemical Name or Material 3-Benzyloxybenzyl alcohol
Assay Percent Range 97%
CAS 1700-30-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00004642
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2OH
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Benzyloxybenzyl alcohol
CAS 1700-30-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H14O2
Linear Formula C6H5CH2OC6H4CH2OH
Formula Weight 214.26
MDL Number MFCD00004642
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents