missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzyloxybenzyl alcohol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC181940010

 View more versions of this product

Catalog No. AC181940010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

214.26
≥96.0%
Light yellow to light beige
96.0
Authentic
97%
C14H14O2
Glass bottle
Powder
1g
47° to 52°C
100.0
1700-30-7
3-Benzyloxybenzyl alcohol
MFCD00004642
C6H5CH2OC6H4CH2OH

Chemical Identifiers

1700-30-7
100.0
C14H14O2
214.26
3-Benzyloxybenzyl alcohol
96.0
97%
C6H5CH2OC6H4CH2OH
MFCD00004642
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.