Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxybenzyl alcohol, 97%

$87.43 - $277.07

Chemical Identifiers

Chemical Name or Material 3-Benzyloxybenzyl alcohol
CAS 1700-30-7
Molecular Formula C14H14O2
Formula Weight 214.26
MDL Number MFCD00004642
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2770706

 
alfa aesar™
H2770706
5g Each for $87.43

AAH2770714

 
alfa aesar™
H2770714
25g Each for $277.07
Description & Specifications

Specifications

Formula Weight 214.26
Chemical Name or Material 3-Benzyloxybenzyl alcohol
Assay 97%
Molecular Formula C14H14O2
Melting Point 47° to 51°C
MDL Number MFCD00004642
CAS 1700-30-7
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.