missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxybenzeneboronic acid, 98+%

$39.64 - $125.23

Chemical Identifiers

CAS 156682-54-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
MDL Number MFCD02093063
InChI Key WIJNYNBSPQMJGO-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
PubChem CID 2734313
IUPAC Name (3-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1747403 View Documents Alfa Aesar™
L1747403
1g Each for $39.64
 
AAL1747406 View Documents Alfa Aesar™
L1747406
5g Each for $125.23
 
Specifications

Chemical Identifiers

156682-54-1
228.054
WIJNYNBSPQMJGO-UHFFFAOYSA-N
2734313
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
C13H13BO3
MFCD02093063
3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
(3-phenylmethoxyphenyl)boronic acid

Specifications

122°C to 124°C
156682-54-1
MFCD02093063
3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
228.054
228.06
3-Benzyloxybenzeneboronic acid
1g
C13H13BO3
6808244
WIJNYNBSPQMJGO-UHFFFAOYSA-N
(3-phenylmethoxyphenyl)boronic acid
2734313
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning