missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxybenzeneboronic acid, 98+%

Manufacturer:  Alfa Aesar™ L1747406

 View more versions of this product

Catalog No. AAL1747406



Specifications

Specifications

3-Benzyloxybenzeneboronic acid
5g
C13H13BO3
6808244
WIJNYNBSPQMJGO-UHFFFAOYSA-N
(3-phenylmethoxyphenyl)boronic acid
2734313
≥98%
122°C to 124°C
156682-54-1
MFCD02093063
3-benzyl-oxy phenyl boranediol
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
228.054
228.06

Chemical Identifiers

156682-54-1
228.054
WIJNYNBSPQMJGO-UHFFFAOYSA-N
2734313
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
C13H13BO3
MFCD02093063
3-benzyl-oxy phenyl boranediol
(3-phenylmethoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents