Please login to your online account to display your discounted pricing

3-Benzyloxybenzaldehyde, 98%, Acros Organics


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  106070250

 View more versions of this product

Catalog No. AC106070250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 212.25
Quantity 25g
Color White to light beige
EINECS Number 216-932-8
TSCA TSCA
Melting Point 55° to 59°C
CAS Max % 100.0
Chemical Name or Material 3-Benzyloxybenzaldehyde
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CHO
MDL Number MFCD00003367
Name Note 98%
Assay Percent Range 98%
Molecular Formula C14H12O2
CAS 1700-37-4
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Benzyloxybenzaldehyde
Name Note 98%
CAS 1700-37-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H12O2
Linear Formula C6H5CH2OC6H4CHO
Formula Weight 212.25
MDL Number MFCD00003367