missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Benzyloxybenzaldehyde, 98%, ACROS Organics™
Chemical Identifiers
CAS | 1700-37-4 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.25 |
MDL Number | MFCD00003367 |
InChI Key | JAICGBJIBWDEIZ-UHFFFAOYSA-N |
Synonym | 3-benzyloxy benzaldehyde, 3-benzyloxy-benzaldehyde, 3-benzyloxybenzaldehyde, 3-phenylmethoxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, benzaldehyde, m-benzyloxy, k1s6ob2gnn, m-benzyloxybenzaldehyde, unii-k1s6ob2gnn |
PubChem CID | 74342 |
IUPAC Name | 3-phenylmethoxybenzaldehyde |
SMILES | C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC106071000
|
Thermo Fisher Scientific
106071000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1700-37-4 | |
212.25 | |
JAICGBJIBWDEIZ-UHFFFAOYSA-N | |
74342 | |
C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O |
C14H12O2 | |
MFCD00003367 | |
3-benzyloxy benzaldehyde, 3-benzyloxy-benzaldehyde, 3-benzyloxybenzaldehyde, 3-phenylmethoxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, benzaldehyde, m-benzyloxy, k1s6ob2gnn, m-benzyloxybenzaldehyde, unii-k1s6ob2gnn | |
3-phenylmethoxybenzaldehyde |
Specifications
Authentic | |
White to Beige | |
100g | |
97.5 | |
98% | |
C6H5CH2OC6H4CHO | |
3-benzyloxy benzaldehyde, 3-benzyloxy-benzaldehyde, 3-benzyloxybenzaldehyde, 3-phenylmethoxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, benzaldehyde, m-benzyloxy, k1s6ob2gnn, m-benzyloxybenzaldehyde, unii-k1s6ob2gnn | |
C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O | |
212.25 | |
212.25 | |
98% |
Glass bottle | |
55°C to 59°C | |
1700-37-4 | |
100.0 | |
C14H12O2 | |
MFCD00003367 | |
JAICGBJIBWDEIZ-UHFFFAOYSA-N | |
3-phenylmethoxybenzaldehyde | |
74342 | |
Fine Crystalline Powder | |
3-Benzyloxybenzaldehyde, 98% |
Safety and Handling
EINECSNumber : 216-932-8
TSCA : TSCA