missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Benzyloxybenzaldehyde, 98%, Thermo Scientific Chemicals
$69.33 - $374.97
Chemical Identifiers
CAS | 1700-37-4 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.248 |
MDL Number | MFCD00003367 |
InChI Key | JAICGBJIBWDEIZ-UHFFFAOYSA-N |
Synonym | 3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn |
PubChem CID | 74342 |
IUPAC Name | 3-phenylmethoxybenzaldehyde |
SMILES | C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2290906
|
Thermo Scientific Chemicals
B2290906 |
5 g |
Each for $69.33
|
|
|||||
AAB2290914
|
Thermo Scientific Chemicals
B2290914 |
25 g |
Each for $131.97
|
|
|||||
AAB2290922
|
Thermo Scientific Chemicals
B2290922 |
100 g |
Each for $374.97
|
|
|||||
Description
3-Benzyloxybenzaldehyde is used as a pharmaceutical intermediate. It activates the adenyl cyclase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Benzyloxybenzaldehyde is used as a pharmaceutical intermediate. It activates the adenyl cyclase.
Solubility
Soluble in water 38.87 mg/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
1700-37-4 | |
212.248 | |
JAICGBJIBWDEIZ-UHFFFAOYSA-N | |
74342 | |
C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O |
C14H12O2 | |
MFCD00003367 | |
3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn | |
3-phenylmethoxybenzaldehyde |
Specifications
1700-37-4 | |
180°C (0.08 mmHg) | |
MFCD00003367 | |
1956589 | |
3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn | |
JAICGBJIBWDEIZ-UHFFFAOYSA-N | |
3-phenylmethoxybenzaldehyde | |
74342 | |
98% |
54°C to 58°C | |
C14H12O2 | |
5 g | |
Air Sensitive | |
Soluble in water 38.87mg/L @ 25°C. | |
C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O | |
212.248 | |
212.25 | |
3-Benzyloxybenzaldehyde |
Safety and Handling
EINECSNumber : 216-932-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only