Please login to your online account to display your discounted pricing

3-Benzyloxyaniline, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3-Benzyloxyaniline
CAS 1484-26-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H13NO
Linear Formula C6H5CH2OC6H4NH2
Formula Weight 199.25
MDL Number MFCD00007784
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146260050

  SDS
thermo fisher scientific
AC146260050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC146260100

  SDS
thermo fisher scientific
AC146260100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 199.25
Physical Form Powder
Quantity 5g
Color Beige to tan
Melting Point 62° to 66°C
Beilstein 13, 404
CAS Max % 100.0
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4NH2
Chemical Name or Material 3-Benzyloxyaniline
Assay Percent Range 98%
CAS 1484-26-0
Molecular Formula C13H13NO
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00007784
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.