missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxyaniline, 98%

Manufacturer:  Alfa Aesar™ H5570803

 View more versions of this product

Catalog No. AAH5570803



Specifications

Specifications

3-Benzyloxyaniline
63°C to 67°C
1484-26-0
MFCD00007784
IGPFOKFDBICQMC-UHFFFAOYSA-N
3-phenylmethoxyaniline
92892
Solid
Brown
1g
C13H13NO
3-benyloxyaniline
C1=CC=C(C=C1)COC2=CC=CC(=C2)N
199.253
199.25
98%

Chemical Identifiers

1484-26-0
199.253
IGPFOKFDBICQMC-UHFFFAOYSA-N
92892
C1=CC=C(C=C1)COC2=CC=CC(=C2)N
C13H13NO
MFCD00007784
3-benyloxyaniline
3-phenylmethoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 216-056-6

TSCA : No

Recommended Storage : Ambient temperatures

Documents