missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzyloxyacetophenone, 97%, ACROS Organics™

$46.45 - $46.45

Chemical Identifiers

CAS 34068-01-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.27
MDL Number MFCD00026221
InChI Key FGQMEAWGAUALJQ-UHFFFAOYSA-N
Synonym 1-3-benzyloxy phenyl ethan-1-one, 1-3-benzyloxy phenyl ethanone, 1-3-phenylmethoxy phenyl ethan-1-one, 1-acetyl-3-phenylmethoxy benzene, 3'-benzyloxy acetophenone, 3'-benzyloxyacetophenone, 3-benzyloxyacetophenone, ethanone, 1-3-phenylmethoxy phenyl
PubChem CID 98689
IUPAC Name 1-(3-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC353120050
SDS View Documents
Thermo Fisher Scientific
353120050
5g Each for $46.45
 
Specifications

Chemical Identifiers

34068-01-4
226.27
FGQMEAWGAUALJQ-UHFFFAOYSA-N
98689
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
C15H14O2
MFCD00026221
1-3-benzyloxy phenyl ethan-1-one, 1-3-benzyloxy phenyl ethanone, 1-3-phenylmethoxy phenyl ethan-1-one, 1-acetyl-3-phenylmethoxy benzene, 3'-benzyloxy acetophenone, 3'-benzyloxyacetophenone, 3-benzyloxyacetophenone, ethanone, 1-3-phenylmethoxy phenyl
1-(3-phenylmethoxyphenyl)ethanone

Specifications

Authentic
Yellow
34068-01-4
100.0
C15H14O2
08, IV, 335
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-(3-phenylmethoxyphenyl)ethanone
98689
Low Melting Solid
3-Benzyloxyacetophenone, 97%
Glass bottle
5g
96.0
96% min. (GC)
MFCD00026221
1-3-benzyloxy phenyl ethan-1-one, 1-3-benzyloxy phenyl ethanone, 1-3-phenylmethoxy phenyl ethan-1-one, 1-acetyl-3-phenylmethoxy benzene, 3'-benzyloxy acetophenone, 3'-benzyloxyacetophenone, 3-benzyloxyacetophenone, ethanone, 1-3-phenylmethoxy phenyl
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
226.27
226.27
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 251-816-0

TSCA : TSCA