missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzyloxybenzeneboronic acid, 98%, Acros Organics™

Chemical Identifiers

CAS 156682-54-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
InChI Key WIJNYNBSPQMJGO-UHFFFAOYSA-N
Synonym 3-benzyl-oxy phenyl boranediol
PubChem CID 2734313
IUPAC Name (3-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC358865000 SDS View Documents Thermo Fisher Scientific
358865000
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

156682-54-1
228.054
3-benzyl-oxy phenyl boranediol
(3-phenylmethoxyphenyl)boronic acid
C13H13BO3
WIJNYNBSPQMJGO-UHFFFAOYSA-N
2734313
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O

Specifications

Authentic
White to Beige
500mg
97.5
97%
3-benzyl-oxy phenyl boranediol
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
228.054
228.05
98%
Glass Bottle
141.0°C to 142.0°C
156682-54-1
100.0
C13H13BO3
WIJNYNBSPQMJGO-UHFFFAOYSA-N
(3-phenylmethoxyphenyl)boronic acid
2734313
Crystalline Powder or Crystals
3-Benzyloxybenzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.