Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxy-4-methoxybenzoic acid, 97+%

$55.55 - $689.59

Chemical Identifiers

Chemical Name or Material 3-Benzyloxy-4-methoxybenzoic acid
CAS 58452-00-9
Molecular Formula C15H14O4
Formula Weight 258.28
MDL Number MFCD00016526
Beilstein 2660886
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0573103

 
alfa aesar™
L0573103
1g Each for $55.55

AAL0573106

 
alfa aesar™
L0573106
5g Each for $178.55

AAL0573114

 
alfa aesar™
L0573114
25g Each for $689.59
Description & Specifications

Specifications

Formula Weight 258.28
Chemical Name or Material 3-Benzyloxy-4-methoxybenzoic acid
Assay ≥97%
Molecular Formula C15H14O4
MDL Number MFCD00016526
CAS 58452-00-9
Beilstein 2660886
Melting Point 177° to 182°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Benzyloxy-4-methoxybenzoic acid is used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.