Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxy-4-methoxybenzaldehyde, 98%

$56.04 - $56.04

Chemical Identifiers

Chemical Name or Material 3-Benzyloxy-4-methoxybenzaldehyde
CAS 6346-05-0
Molecular Formula C15H14O3
Formula Weight 242.27
MDL Number MFCD00003386
Beilstein 794627
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0683009

 
alfa aesar™
L0683009
10g Each for $56.04
Description & Specifications

Specifications

Formula Weight 242.27
Chemical Name or Material 3-Benzyloxy-4-methoxybenzaldehyde
Assay 98%
Molecular Formula C15H14O3
MDL Number MFCD00003386
CAS 6346-05-0
Quantity 10g
Beilstein 794627
Melting Point 60° to 64°C