missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxy-4-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Synonym 3-Benzyloxy-4-fluorophenylboronic acid pinacol ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62335MD View Documents Alfa Aesar™
H62335MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6233503 View Documents Alfa Aesar™
H6233503
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

3-Benzyloxy-4-fluorophenylboronic acid pinacol ester

Specifications

250mg
328.19
96%
3-Benzyloxy-4-fluorophenylboronic acid pinacol ester
Solid
3-Benzyloxy-4-fluorobenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures