missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzyloxy-2-hydroxy-1-propylamine, 99%

$113.69 - $393.43

Chemical Identifiers

CAS 90503-15-4
Molecular Formula C10H15NO2
Formula Weight 181.24
MDL Number MFCD00183222
Synonym 1-Amino-3-benzyloxy-2-propanol
Chemical Name or Material 3-Benzyloxy-2-hydroxy-1-propylamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5024803 SDS Alfa Aesar™
H5024803
1g Each for $393.43
AAH50248MD SDS Alfa Aesar™
H50248MD
250mg Each for $113.69
Specifications Description & Specifications

Chemical Identifiers

90503-15-4
181.24
1-Amino-3-benzyloxy-2-propanol
C10H15NO2
MFCD00183222
3-Benzyloxy-2-hydroxy-1-propylamine

Specifications

90503-15-4
181.24
1-Amino-3-benzyloxy-2-propanol
3-Benzyloxy-2-hydroxy-1-propylamine
C10H15NO2
MFCD00183222
1g
99%