missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzyloxazolidine, TCI America™

$204.72 - $517.92

Chemical Identifiers

CAS 13657-16-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00059028
InChI Key FQNODHUYZYLTPN-UHFFFAOYSA-N
PubChem CID 319258
IUPAC Name 3-benzyl-1,3-oxazolidine
SMILES C1COCN1CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13391G View Documents TCI America
B13391G
1g Each for $204.72
 
B13395G View Documents TCI America
B13395G
5g Each for $517.92
 
Specifications

Chemical Identifiers

13657-16-4
163.22
FQNODHUYZYLTPN-UHFFFAOYSA-N
3-benzyl-1,3-oxazolidine
C10H13NO
MFCD00059028
319258
C1COCN1CC2=CC=CC=C2

Specifications

127°C
1g
C10H13NO
FQNODHUYZYLTPN-UHFFFAOYSA-N
3-benzyl-1,3-oxazolidine
319258
Clear Liquid at 20°C
3-Benzyloxazolidine
Yellow
13657-16-4
MFCD00059028
C1COCN1CC2=CC=CC=C2
163.22
163.22
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No